Novus Biologicals
Manufacturer Code:NBP270217AF750
Catalog # NP270217A70
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B10 |
Immunogen | Synthetic peptide corresponding to residues near C-terminus of human Argininosuccinate Synthase (NP_000041). |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Argininosuccinate synthase; argininosuccinate synthase argininosuccinate synthase 1 argininosuccinate synthetase argininosuccinate synthetase 1 ASSCTLN1 citrulline-aspartate ligase Citrulline--aspartate ligase EC 6.3.4.5; argininosuccinate synthetase 1; Citrulline--aspartate ligase; citrulline-aspartate ligase
Gene Aliases: ASS; ASS1; CTLN1
UniProt ID: (Human) P00966
Entrez Gene ID: (Human) 445
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.