Novus Biologicals
Manufacturer Code:NBP246548
Catalog # NBP246548
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D4 |
Immunogen | Full length human recombinant protein of human ASL (NP_001020114) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: argininosuccinase; argininosuccinase argininosuccinate lyase Arginosuccinase ASAL EC 4.3.2.1; Argininosuccinate lyase; Arginosuccinase; ASAL
Gene Aliases: ASAL; ASL
UniProt ID: (Human) P04424
Entrez Gene ID: (Human) 435
Molecular Function:
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.