Invitrogen
Manufacturer Code:MA531576
Catalog # PIMA531576
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT825 |
Immunogen | Full length human Arginase 1 recombit protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ARG1 antibody Arginase 1 antibody; arginase 1, liver; arginase I; arginase, liver; Arginase-1; Liver-type arginase; Type I arginase
Gene Aliases: AI; AI256583; Arg-1; ARG1; PGIF
UniProt ID: (Human) P05089, (Mouse) Q61176
Entrez Gene ID: (Human) 383, (Mouse) 11846
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.