Novus Biologicals
Manufacturer Code:NBP292835100UL
Catalog # NB027536
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 17-340 of human RARS2 (NP_064716.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: arginine-tRNA ligase arginyl-tRNA synthetase 2 mitochondrial Arginyl-tRNA synthetase-like ArgRS DALRD2 dJ382I10.6 EC 6.1.1.19 MGC14993 MGC23778 PCH6 PRO1992 probable arginyl-tRNA synthetase mitochondrial RARSLArginine--tRNA ligase; Arginyl-tRNA synthetase; ArgRS; Probable arginine--tRNA ligase, mitochondrial; probable arginyl-tRNA synthetase, mitochondrial
Gene Aliases: ArgRS; DALRD2; PCH6; PRO1992; RARS2; RARSL
UniProt ID: (Human) Q5T160
Entrez Gene ID: (Human) 57038
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.