Novus Biologicals
Manufacturer Code:NB10065214
Catalog # NB10065214
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic arginine-vasopressin conjugated to bovine thyroglobulin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antidiuretic hormone; Arg-vasopressin; Arginine-vasopressin; AVP-NPII; Copeptin; neurohypophyseal; Neurophysin 2; Neurophysin-II; prepro-arginine-vasopressin-neurophysin II; prepro-AVP-NP II; Vasopressin-neurophysin 2-copeptin; vasopressin-neurophysin II-copeptin
Gene Aliases: ADH; ARVP; AVP; AVP-NPII; AVRP; VP
UniProt ID: (Human) P01185
Entrez Gene ID: (Human) 551
Molecular Function:
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.