Invitrogen
Manufacturer Code:PA5114872
Catalog # PIPA5114872
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AQP9 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700020I22Rik AI266899 Aqp9 AQP-9 Aquaglyceroporin-9 aquaporin 9 aquaporin-9 HsT17287 neutral solute channel aquaporin 9 small solute channel 1 SSC1 T17287; AQP-9; Aquaglyceroporin-9; Aquaporin-9; neutral solute channel aquaporin 9; Small solute channel 1
Gene Aliases: 1700020I22Rik; AI266899; AQP-9; AQP9; HsT17287; SSC1; T17287
UniProt ID: (Human) O43315, (Mouse) Q9JJJ3, (Rat) P56627
Entrez Gene ID: (Human) 366, (Mouse) 64008, (Rat) 65054
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.