Novus Biologicals
Manufacturer Code:NBP299492DL594
Catalog # NB061185
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Aquaporin-9. |
Conjugate | DyLight 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AQP-9; AQP-9 Aquaglyceroporin-9 aquaporin 9 aquaporin-9 HsT17287 Small solute channel 1 SSC1aquaglyceroporin-9; Aquaglyceroporin-9; Aquaporin-9; Small solute channel 1
Gene Aliases: AQP-9; AQP9; HsT17287; SSC1; T17287
UniProt ID: (Human) O43315
Entrez Gene ID: (Human) 366
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.