Invitrogen
Manufacturer Code:PA536529
Catalog # PIPA536529
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 220-270 of human AQP5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AQP-5; Aquaporin-5; Aquaporin-5 AQP-5 Aquaporin5 AQP5 Aquaporin 5 AQP 5
Gene Aliases: AQP-5; AQP5; PPKB
UniProt ID: (Human) P55064, (Mouse) Q9WTY4, (Rat) P47864
Entrez Gene ID: (Human) 362, (Mouse) 11830, (Rat) 25241
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.