Invitrogen
Manufacturer Code:PA577840
Catalog # PIPA577840
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced against the C-terminal peptide (Sequence N-CHLEQPPPSTEAENVKLAHMKHKEQI) of rat aquaporin 3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 31.4 kDa water channel protein; AQP-3; AQP3 AQP 3 AQP-3 Aqp3 AQP3&lowbarHUMAN Aquaglyceroporin-3 Aquaporin 3 (GIL blood group) Aquaporin 3 (Gill blood group) Aquaporin-3 Aquaporin3 GIL Gill blood group; Aquaglyceroporin-3; aquaporin 3 (GIL blood group); Aquaporin-3
Gene Aliases: AQP-2; AQP-3; AQP3; GIL
UniProt ID: (Human) Q92482, (Rat) P47862, (Mouse) Q8R2N1
Entrez Gene ID: (Human) 360, (Rat) 65133, (Mouse) 11828
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.