Invitrogen
Manufacturer Code:PA5114871
Catalog # PIPA5114871
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AQP3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 31.4 kDa water channel protein; 31.4 kDa water channel protein AQP 3 AQP-2 Aqp3 AQP-3 Aquaglyceroporin-3 aquaporin 3 aquaporin 3 (GIL blood group) aquaporin 3 (Gill blood group) Aquaporin3 aquaporin-3 GIL Gill blood group; AQP-3; Aquaglyceroporin-3; aquaporin 3 (GIL blood group); Aquaporin-3
Gene Aliases: AQP-2; AQP-3; AQP3; GIL
UniProt ID: (Human) Q92482, (Rat) P47862, (Mouse) Q8R2N1
Entrez Gene ID: (Human) 360, (Rat) 65133, (Mouse) 11828
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.