Novus Biologicals
Manufacturer Code:NBP29288520UL
Catalog # NB027631
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 178-292 of human AQP3 (NP_004916.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AQP-3; AQP-3 Aquaglyceroporin-3 aquaporin 3 aquaporin 3 (GIL blood group) aquaporin 3 (Gill blood group) aquaporin-3 GIL Gill blood group; Aquaglyceroporin-3; aquaporin 3 (GIL blood group); Aquaporin-3
Gene Aliases: AQP-3; AQP3; GIL
UniProt ID: (Human) Q92482
Entrez Gene ID: (Human) 360
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.