Invitrogen
Manufacturer Code:PA578809
Catalog # PIPA578809
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human Aquaporin 2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADH water channel; AQP-2; AQP-CD; aquaporin 2 (collecting duct); Aquaporin-2; Aquaporin-2 AQP-2 ADH water channel Aquaporin-CD AQP-CD Collecting duct water channel protein WCH-CD Water channel protein for renal collecting duct AQP2; Aquaporin-CD; Collecting duct water channel protein; Water channel protein for renal collecting duct; water-channel aquaporin 2; WCH-CD
Gene Aliases: AQP-2; AQP-CD; AQP2; aquaporin-2; cph; jpk; WCH-CD
UniProt ID: (Human) P41181, (Mouse) P56402, (Rat) P34080
Entrez Gene ID: (Human) 359, (Mouse) 11827, (Rat) 25386
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.