Invitrogen
Manufacturer Code:PA578457
Catalog # PIPA578457
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of Human Aquaporin 2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADH water channel; AQP CD AQP-2 AQP2 WCH CD; AQP-2; AQP-CD; aquaporin 2 (collecting duct); Aquaporin-2; Aquaporin-CD; Collecting duct water channel protein; Water channel protein for renal collecting duct; water-channel aquaporin 2; WCH-CD
Gene Aliases: AQP-2; AQP-CD; AQP2; aquaporin-2; WCH-CD
UniProt ID: (Human) P41181, (Rat) P34080
Entrez Gene ID: (Human) 359, (Rat) 25386
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.