Invitrogen
Manufacturer Code:PA577715
Catalog # PIPA577715
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)SLSLTKYHFDER corresponding to amino acid residues 140- 151 of rat Aquaporin 11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AQP-11; AQP11; Aquaporin-11; CG12251 gene product
Gene Aliases: 1700015P13Rik; AI648923; Aqp11; sjds
UniProt ID: (Rat) Q8CHM1, (Mouse) Q8BHH1
Entrez Gene ID: (Rat) 286758, (Mouse) 66333
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.