Invitrogen
Manufacturer Code:MA535461
Catalog # PIMA535461
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0925 |
Immunogen | A synthesized peptide derived from human AQP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AQP 1 AQP CHIP Aqp1 AQP-1 AQP-CHIP Aquaporin CHIP Aquaporin1 Aquaporin-1 Aquaporin-CHIP CHIP 28 Chip28 CO Colton blood group Delayed early response protein 2 DER2 MGC26324 Urine water channel Water channel protein for red blood cells and kidney proximal tubule; AQP-1; Aquaporin 1 (aquaporin channel forming integral protein 28 (CHIP)); Aquaporin-1; Aquaporin-CHIP; Delayed early response protein 2; DER2; growth factor-induced delayed early response protein; Water channel protein for red blood cells and kidney proximal tubule
Gene Aliases: Aqp1; CHIP28
UniProt ID: (Mouse) Q02013, (Rat) P29975
Entrez Gene ID: (Mouse) 11826, (Rat) 25240
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.