Abnova
Manufacturer Code:PAB28846
Catalog # 89-130-369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to Aqp1 N-terminus. |
Conjugate | ATTO 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant Rat Aqp1.
For Research Use Only
Protein Aliases: AQP-1; Aquaporin 1 (aquaporin channel forming integral protein 28 (CHIP)); Aquaporin-1; Aquaporin-CHIP; CHIP28; Water channel protein for red blood cells and kidney proximal tubule
Gene Aliases: Aqp1; CHIP28
UniProt ID: (Rat) P29975
Entrez Gene ID: (Rat) 25240
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.