Novus Biologicals
Manufacturer Code:NBP298661AF700
Catalog # NB098346
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human Aprataxin fragment. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AOA1EAOH AOAMGC1072 aprataxin ataxia 1 early onset with hypoalbuminemia AXA1 EC 3.- EOAHA FHA-HIT FLJ20157 Forkhead-associated domain histidine triad-like protein; Aprataxin; FHA-HIT; Forkhead-associated domain histidine triad-like protein
Gene Aliases: AOA; AOA1; APTX; AXA1; EAOH; EOAHA; FHA-HIT
UniProt ID: (Human) Q7Z2E3
Entrez Gene ID: (Human) 54840
Molecular Function:
DNA binding protein
damaged DNA-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.