Novus Biologicals
Manufacturer Code:NBP267161
Catalog # NBP267161
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JF0548 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Apo-AI; APOA1 Apo-AI ApoA-I Apolipoprotein A1 apolipoprotein A-I MGC117399; Apolipoprotein A-I; Apolipoprotein A-I(1-242); Apolipoprotein A1; ProapoA-I; Proapolipoprotein A-I; Truncated apolipoprotein A-I
Gene Aliases: apo(a); APOA1
UniProt ID: (Human) P02647
Entrez Gene ID: (Human) 335
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.