Invitrogen
Manufacturer Code:MA530323
Catalog # PIMA530323
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 102 |
Immunogen | Recombit Mouse Clusterin/Apolipoprotein J/Apo-J Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Apo-J; Apolipoprotein J; Clusterin; Clusterin alpha chain; Clusterin beta chain; Clustrin; complement lysis inhibitor; SGP-2; Sulfated glycoprotein 2; testosterone repressed prostate message; testosterone repressed prostate message-2
Gene Aliases: AI893575; ApoJ; Cli; Clu; D14Ucla3; Msgp-2; Sgp-2; Sgp2; SP-40; Sugp-2
UniProt ID: (Mouse) Q06890
Entrez Gene ID: (Mouse) 12759
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.