Invitrogen
Manufacturer Code:PA546931
Catalog # PIPA546931
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Clusterin Glu22-Glu448 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Apo-J; APO-J APOJ CLI KUB1 NA1/NA2 SGP-2 SGP2 SP-40 TRPM-2 TRPM2 aging-associated protein 4 apolipoprotein J complement cytolysis inhibitor complement lysis inhibitor complement-associated protein SP-4040 ku70-binding protein 1 sulfated glycoprotein 2 testosterone-repressed prostate message 2; Apolipoprotein J; Clusterin; Clusterin alpha chain; Clusterin beta chain; Clustrin; complement lysis inhibitor; SGP-2; Sulfated glycoprotein 2; testosterone repressed prostate message; testosterone repressed prostate message-2
Gene Aliases: AI893575; ApoJ; Cli; Clu; D14Ucla3; Msgp-2; Sgp-2; Sgp2; SP-40; Sugp-2
UniProt ID: (Mouse) Q06890
Entrez Gene ID: (Mouse) 12759
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.