Invitrogen
Manufacturer Code:PA5110616
Catalog # PIPA5110616
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human Apolipoprotein J |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAG4 Aging-associated gene 4 protein AI893575 Apo J a Apo J alpha Apo J b Apo J beta Apo J a Apo J Apo Ja Apo Jb Apoj Apo-J ApoJ a ApoJ alpha ApoJ b ApoJ beta ApoJ a ApoJ ApoJa ApoJalpha ApoJb ApoJbeta Apolipoprotein J CLI CLU CLU1 CLU2 Clusterin Clusterin alpha chain Clusterin beta chain Clustrin Complement cytolysis inhibitor Complement cytolysis inhibitor a chain Complement cytolysis inhibitor b chain Complement-associated protein SP-4040 D14Ucla3 DAG Dimeric acid glycoprotein HGNC:2095 Ku70-binding protein 1 KUB1 MGC24903 Msgp-2 NA1/NA2 RATTRPM2B Sgp2 SGP-2 SP40 SP-40 Sugp-2 Sulfated glycoprotein 2 Testosterone repressed prostate message 2 Testosterone-repressed prostate message 2 TRPM2 TRPM-2 TRPM2B Trpmb; Aging-associated gene 4 protein; aging-associated protein 4; Apo-J; ApoJalpha; ApoJbeta; Apolipoprotein J; CLI; Clusterin; Clusterin alpha chain; Clusterin beta chain; Complement cytolysis inhibitor; Complement cytolysis inhibitor a chain; Complement cytolysis inhibitor b chain; complement lysis inhibitor; Complement-associated protein SP-40,40; Ku70-binding protein 1; NA1/NA2; SGP-2; Sulfated glycoprotein 2; Testosterone-repressed prostate message 2; TRPM-2
Gene Aliases: AAG4; APO-J; APOJ; CLI; CLU; CLU1; CLU2; KUB1; NA1/NA2; SGP-2; SGP2; SP-40; TRPM-2; TRPM2
UniProt ID: (Human) P10909
Entrez Gene ID: (Human) 1191
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.