Invitrogen
Manufacturer Code:PA5114866
Catalog # PIPA5114866
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human APOH corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2GPI Activated protein C-binding protein Anticardiolipin cofactor APC inhibitor Apoh Apo-H Apolipoprotein H apolipoprotein H (beta-2-glycoprotein I) B2G1 B2gp1 B2GPI beta(2)GPI beta-2-glycoprotein 1 Beta-2-glycoprotein I beta-2-GPI beta2-GPI BG; Activated protein C-binding protein; Anticardiolipin cofactor; APC inhibitor; Apo-H; Apolipoprotein H; apolipoprotein H (beta-2-glycoprotein I); B2GPI; beta(2)GPI; Beta-2-glycoprotein 1; Beta-2-glycoprotein I
Gene Aliases: APOH; B2G1; B2GP1; B2GPI; beta-2-GPI; beta2-GPI; BG
UniProt ID: (Human) P02749, (Mouse) Q01339
Entrez Gene ID: (Human) 350, (Rat) 287774, (Mouse) 11818
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.