Novus Biologicals
Manufacturer Code:NB1002217AF594
Catalog # N102217A594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to a C-terminal portion of the human ApoER2 protein sequence (between residues 800-900). [UniProtNo. Q14114] |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
Protein Aliases: ApoE receptor 2; APOER2HSZ75190 Apolipoprotein E receptor 2 low density lipoprotein receptor-related protein 8 apolipoprotein e receptor low-density lipoprotein receptor-related protein 8 LRP-8 MCI1; Apolipoprotein E receptor 2; low density lipoprotein receptor-related protein 8, apolipoprotein e receptor; Low-density lipoprotein receptor-related protein 8; LRP-8
Gene Aliases: APOER2; HSZ75190; LRP-8; LRP8; MCI1
UniProt ID: (Human) Q14114
Entrez Gene ID: (Human) 7804
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.