Novus Biologicals
Manufacturer Code:NBP222110SS
Catalog # NBP222110SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1H4 |
Immunogen | Purified recombinant fragment of human ApoE expressed in E. coli. [UniProt# P02649] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Apo-E; Apo-E apolipoprotein E apolipoprotein E3 late onset) MGC1571; Apolipoprotein E; apolipoprotein E3
Gene Aliases: AD2; APO-E; APOE; LDLCQ5; LPG
UniProt ID: (Human) P02649
Entrez Gene ID: (Human) 348
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.