Novus Biologicals
Manufacturer Code:NBP241216
Catalog # NBP241216
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human APO-E. The immunogen is located within amino acids 220 - 270 of APO-E. Amino Acid Squence: ARMEEMGSRTRDRLDEVKE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apo-E; Apo-E apolipoprotein E apolipoprotein E3 late onset) MGC1571; Apolipoprotein E; apolipoprotein E3
Gene Aliases: AD2; APO-E; APOE; LDLCQ5; LPG
UniProt ID: (Human) P02649
Entrez Gene ID: (Human) 348
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.