Invitrogen
Manufacturer Code:PA537838
Catalog # PIPA537838
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-ENGKIKVLNQE from the internal region of APOD (aa70-80) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Apo-D; APOD apo-D; Apolipoprotein D
Gene Aliases: APOD
UniProt ID: (Human) P05090
Entrez Gene ID: (Human) 347
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.