Novus Biologicals
Manufacturer Code:NBP292734100UL
Catalog # NB027337
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-99 of human APOC3 (NP_000031.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apo-CIII; apoC-III; APOC3 apo-CIII ApoC-III APOCIII Apolipoprotein C3 apolipoprotein C-III MGC150353; Apolipoprotein C-III; Apolipoprotein C3
Gene Aliases: APOC3; APOCIII; HALP2
UniProt ID: (Human) P02656
Entrez Gene ID: (Human) 345
Molecular Function:
apolipoprotein
signaling molecule
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.