Novus Biologicals
Manufacturer Code:NBP280430
Catalog # NB013502
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein with Human APOC3 aa 1-100. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apo-CIII; apoC-III; APOC3 apo-CIII ApoC-III APOCIII Apolipoprotein C3 apolipoprotein C-III MGC150353; Apolipoprotein C-III; Apolipoprotein C3
Gene Aliases: APOC3; APOCIII; HALP2
UniProt ID: (Human) P02656
Entrez Gene ID: (Human) 345
Molecular Function:
apolipoprotein
signaling molecule
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.