Invitrogen
Manufacturer Code:PA122560
Catalog # PIPA122560
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Purified full length synthetic apolipoprotein type C-I. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Apo-CI; Apolipoprotein C-I; apolipoprotein C-I variant I; apolipoprotein C-I variant II; Apolipoprotein C1; Truncated apolipoprotein C-I
Gene Aliases: Apo-CI; apo-CIB; ApoC-I; apoC-IB; APOC1
UniProt ID: (Human) P02654
Entrez Gene ID: (Human) 341
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.