R&D Systems
Manufacturer Code:MAB4497100
Catalog # MAB4497100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2554A |
Immunogen | <i>E. coli</i>-derived human Apolipoprotein C-II/ApoC2 Thr23-Glu101 Accession &num P02655 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: APC2 APOC2 Apo-CII ApoC-II Apolipoprotein C2 apolipoprotein C-II Apolipoprotein CII MGC75082; Apo-CII; Apolipoprotein C-II; Apolipoprotein C2; ProapoC-II; Proapolipoprotein C-II
Gene Aliases: APC2; APO-CII; APOC-II; APOC2
UniProt ID: (Human) P02655
Entrez Gene ID: (Human) 344
Molecular Function:
apolipoprotein
hydrolase
lipase
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.