Novus Biologicals
Manufacturer Code:NBP237598
Catalog # NBP237598
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6G6 |
Immunogen | Purified recombinant fragment of human ApoB expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Apo B-100; Apo B-100 apoB-100 apoB-48 apolipoprotein B (including Ag(x) antigen) apolipoprotein B-100 apolipoprotein B48 FLDB LDLCQ4 mutant Apo B 100; Apo B-48; apolipoprotein B (including Ag(x) antigen); Apolipoprotein B-100; Apolipoprotein B-48; apolipoprotein B48
Gene Aliases: APOB; apoB-100; apoB-48; FLDB; LDLCQ4
UniProt ID: (Human) P04114
Entrez Gene ID: (Human) 338
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.