Novus Biologicals
Manufacturer Code:NB200527
Catalog # NB200527
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse low density lipoprotein (LDL) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -70C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apo B-100; Apo B-100 apoB-100 apoB-48 apolipoprotein B (including Ag(x) antigen) apolipoprotein B-100 apolipoprotein B48 FLDB LDLCQ4 mutant Apo B 100; Apo B-48; apolipoprotein B (including Ag(x) antigen); Apolipoprotein B-100; Apolipoprotein B-48; apolipoprotein B48
Gene Aliases: APOB; apoB-100; apoB-48; FLDB; LDLCQ4
UniProt ID: (Human) P04114
Entrez Gene ID: (Human) 338
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.