Novus Biologicals
Manufacturer Code:NB400139SS
Catalog # NB400139SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G5G9 |
Immunogen | Ni-NTA purified full-length recombinant human Apolipoprotein A5 expressed in E. Coli strain BL21 (DE3). [UniProt# Q6Q788] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Apo-AV; apo-AV APOAV APOA-V Apolipoprotein A5 apolipoprotein A-V FLJ97995 MGC126836 RAP3MGC126838 Regeneration-associated protein 3; Apolipoprotein A-V; apolipoprotein A-V precursor variant 3; Apolipoprotein A5; Regeneration-associated protein 3
Gene Aliases: APOA5; APOAV; RAP3; UNQ411/PRO773
UniProt ID: (Human) Q6Q788
Entrez Gene ID: (Human) 116519
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.