Invitrogen
Manufacturer Code:PA521165
Catalog # PIPA521165
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide near the carboxy terminus of human ApoA4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apo-AIV; apo-AIV apoA-IV apolipoprotein A4; apoA-IV; Apolipoprotein A-IV; Apolipoprotein A4
Gene Aliases: APOA4; APOAIV
UniProt ID: (Human) P06727
Entrez Gene ID: (Chicken) 395780, (Human) 337
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.