R&D Systems
Manufacturer Code:AF8125
Catalog # AF8125
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-dervied recombinant human Apolipoprotein A-IV/ApoA Glu21-Ser396 Accession # P06727 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Apo-AIV; apo-AIV ApoA-IV Apolipoprotein A4 apolipoprotein A-IV Apolipoprotein AIV MGC142154 MGC142156; apoA-IV; Apolipoprotein A-IV; Apolipoprotein A4
Gene Aliases: APOA4
UniProt ID: (Human) P06727
Entrez Gene ID: (Human) 337
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.