Novus Biologicals
Manufacturer Code:NBP275410
Catalog # NBP275410
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JB70-35 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Apo-AII; APOA2 apoAII Apo-AII ApoA-II Apolipoprotein A2 apolipoprotein A-II; Apolipoprotein A-II; Apolipoprotein A-II(1-76); Apolipoprotein A2; ProapoA-II; Proapolipoprotein A-II; Truncated apolipoprotein A-II
Gene Aliases: Apo-AII; ApoA-II; APOA2; apoAII
UniProt ID: (Human) P02652
Entrez Gene ID: (Human) 336
Molecular Function: apolipoprotein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.