R&D Systems
Manufacturer Code:MAB4215
Catalog # MAB4215
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 395906 |
Immunogen | <EM>E. coli</EM>-derived recombinant human ApoA2 Gln24-Gln100 Accession # P02652 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Alp-2 APOA2 apoAII Apo-AII ApoA-II Apolipoprotein A2 apolipoprotein A-II Apolipoprotein AII Hdl-1; Apo-AII; Apolipoprotein A-II; Apolipoprotein A-II(1-76); Apolipoprotein A2; ProapoA-II; Proapolipoprotein A-II; Truncated apolipoprotein A-II
Gene Aliases: Apo-AII; ApoA-II; APOA2; apoAII
UniProt ID: (Human) P02652
Entrez Gene ID: (Human) 336
Molecular Function:
apolipoprotein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.