R&D Systems
Manufacturer Code:MAB36641SP
Catalog # MAB36641SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2083A |
Immunogen | <em>E. coli</em>-derived recombinant human ApoA1 Asp25-Gln267 Accession # P02647 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Alp-1 APOA1 Apo-AI ApoA-I Apolipoprotein A1 apolipoprotein A-I Apolipoprotein AI Brp-14 Ltw-1 Lvtw-1 MGC117399 Sep-1 Sep-2; Apo-AI; Apolipoprotein A-I; Apolipoprotein A-I(1-242); Apolipoprotein A1; ProapoA-I; Proapolipoprotein A-I; Truncated apolipoprotein A-I
Gene Aliases: apo(a); APOA1
UniProt ID: (Human) P02647
Entrez Gene ID: (Human) 335
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.