Invitrogen
Manufacturer Code:PA511429
Catalog # PIPA511429
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 7-36 amino acids from the N-terminal region of human Apobec1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: apolipoprotein B editing complex 1; apolipoprotein B mRNA editing enzyme catalytic polypeptide 1; apolipoprotein B mRNA editing enzyme complex-1; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 1; Apolipoprotein B mRNA-editing enzyme 1; C->U-editing enzyme APOBEC-1; HEPR; mRNA(cytosine(6666)) deaminase 1
Gene Aliases: APOBEC-1; APOBEC1; BEDP; CDAR1; HEPR
UniProt ID: (Human) P41238, (Mouse) P51908
Entrez Gene ID: (Human) 339, (Mouse) 11810
Molecular Function:
deaminase
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.