Invitrogen
Manufacturer Code:PA578802
Catalog # PIPA578802
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human Apolipoprotein CIII recombit protein (Position: S21-A99). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apo-CIII; apoC-III; apolipoprotein C-3; Apolipoprotein C-III; Apolipoprotein C-III Apo-CIII ApoC-III Apolipoprotein C3 APOC3; Apolipoprotein C3
Gene Aliases: apo-CIII; ApoC-III; APOC3; APOCIII; HALP2
UniProt ID: (Human) P02656, (Mouse) P33622
Entrez Gene ID: (Human) 345, (Mouse) 11814, (Rat) 24207
Molecular Function:
apolipoprotein
signaling molecule
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.