Invitrogen
Manufacturer Code:701236
Catalog # PI701236
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 43H22L4 |
Immunogen | Peptide corresponding to amino acids 6071 of human Apo A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Apo-AII; Apolipoprotein A-II; Apolipoprotein A-II Apo-AII ApoA-II; Apolipoprotein A-II(1-76); Apolipoprotein A2; ProapoA-II; Proapolipoprotein A-II; Truncated apolipoprotein A-II
Gene Aliases: Apo-AII; ApoA-II; APOA2; APOAII
UniProt ID: (Human) P02652, (Rat) P04638
Entrez Gene ID: (Human) 336, (Rat) 25649
Molecular Function:
apolipoprotein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.