Invitrogen
Manufacturer Code:PA5114863
Catalog # PIPA5114863
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human APOA2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alp-2 APO A2 APOA2 Apoa-2 ApoAII Apo-AII ApoA-II Apolipoprotein Apolipoprotein A2 Apolipoprotein AII Apolipoprotein A-II Apolipoprotein A-II(1-76) Hdl-1 ProapoA-II Proapolipoprotein A-II Truncated apolipoprotein A-II; Apo-AII; Apolipoprotein A-II; Apolipoprotein A-II(1-76); Apolipoprotein A2; ProapoA-II; Proapolipoprotein A-II; Truncated apolipoprotein A-II
Gene Aliases: Alp-2; Apo-AII; Apoa-2; ApoA-II; APOA2; APOAII; Hdl-1
UniProt ID: (Human) P02652, (Rat) P04638, (Mouse) P09813
Entrez Gene ID: (Human) 336, (Rat) 25649, (Mouse) 11807
Molecular Function:
apolipoprotein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.