Invitrogen
Manufacturer Code:MIA1405
Catalog # PIMIA1405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Radioimmune Assays (RIA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 532 |
Immunogen | Purified human plasma Apo A-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Apo-AI; apo-AI apoA-I; Apolipoprotein A-I; Apolipoprotein A-I(1-242); Apolipoprotein A1; ProapoA-I; Proapolipoprotein A-I; Truncated apolipoprotein A-I
Gene Aliases: Alp-1; apo(a); apo-AI; Apoa-1; apoA-I; APOA1; Brp-14; Ltw-1; Lvtw-1; Sep-1; Sep-2; Sep2
UniProt ID: (Human) P02647, (Mouse) Q00623
Entrez Gene ID: (Human) 335, (Mouse) 11806
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.