Invitrogen
Manufacturer Code:701239
Catalog # PI701239
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 19H20L19 |
Immunogen | Peptide corresponding to amino acids 127142 of human Apolipoprotein A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Alp-1 Apoa-1 Brp-14 Ltw-1 Lvtw-1 APO I Apolipoprotein AI; Apo-AI; apolipoprotein A-1; Apolipoprotein A-I; Apolipoprotein A-I(1-242); Apolipoprotein A1; preproapolipoprotein A-I; ProapoA-I; Proapolipoprotein A-I; Truncated apolipoprotein A-I
Gene Aliases: apo(a); apoA-I; APOA1
UniProt ID: (Human) P02647, (Rat) P04639
Entrez Gene ID: (Human) 335, (Rat) 25081
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.