Novus Biologicals
Manufacturer Code:NBP231176
Catalog # NBP231176
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide derived from C terminal region of human Apelin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGTRL1 ligand; AGTRL1 ligand APEL apelin apelin AGTRL1 ligand APJ endogenous ligand XNPEP2; Apelin; Apelin-13; Apelin-28; Apelin-31; Apelin-36; APJ endogenous ligand
Gene Aliases: APEL; APLN; XNPEP2
UniProt ID: (Human) Q9ULZ1
Entrez Gene ID: (Human) 8862
Molecular Function:
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.