Proteintech
Manufacturer Code:114971AP150UL
Catalog # 50-172-7211
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Apelin Fusion Protein Ag2047 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGTRL1 ligand; APEL apelin APJ endogenous ligand APLN XNPEP2; Apelin; apelin, AGTRL1 ligand; Apelin-13; Apelin-28; Apelin-31; Apelin-36; APJ endogenous ligand; preproapelin
Gene Aliases: 6030430G11Rik; APEL; APLN; XNPEP2
UniProt ID: (Human) Q9ULZ1, (Mouse) Q9R0R4, (Rat) Q9R0R3
Entrez Gene ID: (Human) 8862, (Mouse) 30878, (Rat) 58812
Molecular Function:
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.