Novus Biologicals
Manufacturer Code:NBP297192AF700
Catalog # NB101314
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human Antizyme inhibitor 1 fragment. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antizyme inhibitor 1; antizyme inhibitor 1 AZI OAZIMGC691 OAZINEC 4.1.1.17 ODC1L Ornithine decarboxylase antizyme inhibitorMGC3832; AZI; Ornithine decarboxylase antizyme inhibitor
Gene Aliases: AZI; AZIA1; AZIN1; OAZI; OAZIN; ODC1L
UniProt ID: (Human) O14977
Entrez Gene ID: (Human) 51582
Molecular Function:
decarboxylase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.