Novus Biologicals
Manufacturer Code:NBP214298
Catalog # NBP214298
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: PTTIPVTTRIRIVNFVVMNNKTSAGETFEDLMKDGVFEARFPLHKGEGRL KKTWARWRHMFREQPVDEIRNY |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: anoctamin 9 anoctamin-9 PIG5p53-induced gene 5 protein TMEM16J Transmembrane protein 16JTP53I5 tumor protein p53 inducible protein 5 Tumor protein p53-inducible protein 5; Anoctamin-9; p53-induced gene 5 protein; Transmembrane protein 16J; tumor protein p53 inducible protein 5; Tumor protein p53-inducible protein 5
Gene Aliases: ANO9; PIG5; TMEM16J; TP53I5
UniProt ID: (Human) A1A5B4
Entrez Gene ID: (Human) 338440
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.