Proteintech
Manufacturer Code:101542AP150UL
Catalog # 50-172-6066
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Annexin VII Fusion Protein Ag0206 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Annexin 7 annexin A7 Annexin VII Annexin7 ANX7 ANXA7 SNX SYNEXIN; Annexin A7; Annexin VII; Annexin-7; Synexin
Gene Aliases: AI265384; AI316497; ANX7; ANXA7; OK/SW-cl.95; SNX; SYNEXIN
UniProt ID: (Human) P20073, (Mouse) Q07076
Entrez Gene ID: (Human) 310, (Mouse) 11750
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.