Proteintech
Manufacturer Code:662451IG150UL
Catalog # 50-173-6678
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E6A8 |
Immunogen | Annexin V Fusion Protein Ag1538 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Anchorin CII; Anchorin CII Annexin 5 annexin A5 Annexin V ANX5 ANXA5 Calphobindin I CBP I Endonexin II ENX2 Lipocortin V PAP I PP4 Thromboplastin inhibitor VAC alpha Vascular anticoagulant alpha; Annexin A5; Annexin V; Annexin-5; Calphobindin I; CBP-I; Endonexin II; epididymis secretory protein Li 7; Lipocortin V; PAP-I; Placental anticoagulant protein 4; Placental anticoagulant protein I; PP4; Thromboplastin inhibitor; VAC-alpha; Vascular anticoagulant-alpha
Gene Aliases: ANX5; ANXA5; ENX2; HEL-S-7; PP4; R74653; RPRGL3
UniProt ID: (Human) P08758, (Mouse) P48036
Entrez Gene ID: (Human) 308, (Mouse) 11747
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.